Alfa Aesar™ Boron trifluoride, 99+%

Manufacturer: Alfa Aesar™ 2287414

Quantity 25g

 View more versions of this product

Catalog No. AA2287414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7637-07-2
    Formula Weight 67.81
    MDL Number MFCD00011316
    UN Number UN1008
    Merck Index 14,1349
    Synonym Boron fluoride
    Chemical Name or Material Boron trifluoride

    Specifications

    Formula Weight 67.81
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310-P302+P352-P403
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.
    Store in a well-ventilated place.
    Health Hazard 2 GHS H Statement
    H330-H314-H318
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Odor Pungent
    Physical Form Gas
    MDL Number MFCD00011316
    CAS 7637-07-2
    UN Number UN1008
    Synonym Boron fluoride
    Color Colorless
    DOT Information Transport Hazard Class: 2.3; Proper Shipping Name: BORON TRIFLUORIDE
    RTECS Number ED2275000
    Sensitivity Moisture Sensitive
    Merck Index 14,1349
    Boiling Point −100°C
    Melting Point −127°C
    Health Hazard 1 Danger
    Density 3.076g/mL
    TSCA TSCA
    Quantity 25g
    Chemical Name or Material Boron trifluoride
    Assay ≥99%

    GHS H Statement
    H330-H314-H318
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P305+P351+P338-P309-P310-P302+P352-P403
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN: Wash with plenty of soap and water.
    Store in a well-ventilated place.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit