Learn More
Boron trifluoride-acetic acid complex,35-36.5% BF3, Thermo Scientific Chemicals
Description
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used as a desilylating agent. It is also been used in the protodesilylation of the phenyl group as part of the conversion of the dimethylphenylsilyl group into a hydroxyl group with retention of configuration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff. It is also used as a desilylating agent. It is also been used in the protodesilylation of the phenyl group as part of the conversion of the dimethylphenylsilyl group into a hydroxyl group with retention of configuration.
Solubility
It reacts with water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. Stable, but moisture sensitive. Incompatible with water, moist air, metals, strong oxidizing agents. Store under dry atmosphere.
Specifications
Melting Point | -47°C |
Density | 1.353 |
pH | <1 |
Boiling Point | 143°C to 146°C |
Flash Point | 83°C (181°F) |
Odor | Pungent |
Refractive Index | 1.367 |
Quantity | 1000 mL |
UN Number | UN1742 |
Beilstein | 3686177 |
Show More |
Safety and Handling
Hazard Category | H227-H302-H314 |
Hazard Statement | GHS H Statement H301-H330-H314-H318-H227 Toxic if swallowed. Fatal if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. Combustible liquid. |
Precautionary Statement | P210-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: BORON TRIFLUORIDE ACETIC ACID COMPLEX, LIQUID |
EINECSNumber | 206-768-5 |
RTECSNumber | ED2280000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.