Alfa Aesar™ Boron trifluoride diethylether complex

Manufacturer: Alfa Aesar™ 89089A1

Quantity 1kg

 View more versions of this product

Catalog No. AA89089A1

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 109-63-7
    Molecular Formula BF3(C2H5)2O
    Formula Weight 141.93
    MDL Number MFCD00013194
    UN Number UN2604
    Synonym Boron trifluoride diethyl etherate
    Chemical Name or Material Boron trifluoride diethylether complex

    Specifications

    Formula Weight 141.93
    Health Hazard 3 GHS P Statement
    P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Acrid
    Physical Form Liquid
    Molecular Formula BF3(C2H5)2O
    MDL Number MFCD00013194
    CAS 109-63-7
    UN Number UN2604
    Boiling Point 125° to 126°C
    Quantity 1kg
    Health Hazard 2 GHS H Statement
    H330-H314-H318-H226
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.
    DOT Information Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: BORON TRIFLUORIDE DIETHYL ETHERATE
    TSCA TSCA
    Refractive Index 1.348
    Synonym Boron trifluoride diethyl etherate
    Melting Point −60.4°C
    Health Hazard 1 Danger
    Density 1.125g/mL
    Chemical Name or Material Boron trifluoride diethylether complex

    GHS H Statement
    H330-H314-H318-H226
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.

    GHS P Statement
    P210-P303+P361+P353-P304+P340-P305+P351+P338-P320-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit