Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boron trifluoride dihydrate, 65% BF3, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
Packaging:
Glass bottle
13319-75-0
BF3·2H2O
103.85
MFCD00149901
MJCYPBSRKLJZTB-UHFFFAOYSA-N
boron trifluoride dihydrate,boron fluoride dihydrate,borane, trifluoro-, dihydrate,trifluoroborane dihydrate,bf3.2h2o,bf3 dihydrate,bortrifluorid dihydrat,borontrifluoride dihydrate,dsstox_cid_13264,dsstox_rid_79058
61591
trifluoroborane;dihydrate
B(F)(F)F.O.O
13319-75-0
BF3·2H2O
103.85
MFCD00149901
MJCYPBSRKLJZTB-UHFFFAOYSA-N
boron trifluoride dihydrate,boron fluoride dihydrate,borane, trifluoro-, dihydrate,trifluoroborane dihydrate,bf3.2h2o,bf3 dihydrate,bortrifluorid dihydrat,borontrifluoride dihydrate,dsstox_cid_13264,dsstox_rid_79058
61591
trifluoroborane;dihydrate
B(F)(F)F.O.O
Lewis acid
Assay Percent Range 65% BF3
Linear Formula BF3·2H2O
Solubility Information Solubility in water: miscible
Formula Weight 103.85
Physical Form Liquid
Percent Purity 65.0 to 66.0%
Density 1.6420g/mL
Identification Confirmed
Packaging Glass bottle
Specific Gravity 1.642
Viscosity 7 mPa.s (20°C)
Boiling Point 58°C to 60°C (1.5mmHg)
Color Colorless to Yellow
Melting Point 6°C
Quantity 500mL
Chemical Name or Material Boron trifluoride dihydrate
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger