Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Boron trifluoride-dimethyl sulfide complex, purified, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer:  Alfa Aesar™ 42970AP

Quantity 500mL

 View more versions of this product

Catalog No. AA42970AP


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

353-43-5
129.94
UN2920
BF3·S(CH3)2
MFCD00013193
Boron trifluoride-dimethyl sulfide complex, Purified

Specifications

353-43-5
BF3·S(CH3)2
MFCD00013193
1.235g/mL
Moisture Sensitive
Boron trifluoride-dimethyl sulfide complex, Purified
Liquid
129.94
UN2920
ChemSeal™ bottles
500mL

Safety and Handling

SDS & Safety Documents

GHS H Statement
H224-H314-H318-H335
Extremely flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.

storageNote1 :

  • Packaged under Argon

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit