Learn More
Bellco Glass, Inc. Borosil Funnels, Separatory, Pear-Shaped, PTFE Stopcock, 125mL, 19/26
SDP
Supplier:  Bellco Glass, Inc. 452800841
Encompass_Preferred
Borosil Glass Separating Funnel Pear Shape with stem PTFE Stopcock I/C Glass stopper, USP Type I Borosilicate 3.3, ISO 4800 CS/10 250 mLSpecificationsMaterial: GlassFunnel Cup Volume: 250mLGlass Stopper: PTFEOverall Dimension: 22.44" L x 22.44" W x 16.54" HBorosilicate Glass 3.3Graduation Range: Type I
Catalog No. 50-254-9921