Learn More
Bellco Glass, Inc. Borosil Funnels, Separatory, Pear-Shaped, Boroflo PTFE Screw Thread, 250mL, 24/29
SDP
Supplier:  Bellco Glass, Inc. 452800837
Encompass_Preferred
Borosil Glass Separating Funnel, Pear Shape, Fitted with Boroflo Stopcock with PTFE Key and Interchangeable Glass Stopper, 250 mL, CS/10These Separating Funnels, Pear Shaped with a stem, are made of 3.3 borosilicate glass. Complies with IS 1575, DIN/ ISO 4800. Generally used in the separation phase, they come equipped with a Boroflo stopcock with PTFE key. Material: 3.3 Borosilicate GlassColor: ClearCapacity: 250 mLClass/ Quality Grade: Type IStopper Size: 24 / 29Quantity per Case: 10
Catalog No. 50-254-9925