Learn More
Bellco Glass, Inc. Borosil Tubes, Culture, Round Bottom, Clear, PTFE-Lined PP Screw Caps, 50mL
SDP
Supplier:  Bellco Glass, Inc. 452800887
Encompass_Preferred
Borosil Culture Tubes Media Round Bottom PP cap with Liner 50 mL Borosilicate CS/50SpecificationsMaterial: 3.3 Borosilicate Glass Tube size: 50 mLColor: ClearCap Material: Polypropylene Overall Dimension: 9900012Class/Quality Grade: Type I, Class A
Catalog No. 50-254-9875