Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Grade GF/A Filter for Personal Air Samplers

These aerosol sampling and particulate monitoring filters provide high flow rates and minimal sample interference. Cytiva Whatman Grade GF/A card-mounted filters are used in static sample and personal air sampler applications.

Supplier:  Cytiva 1820900086

Catalog No. 09-920-095


Add to Cart

Description

Description

  • Heat resistant (up to 500°C)
  • Made of high-quality borosilicate glass fibers without added binders
  • Low moisture affinity and high loading capacity
Specifications

Specifications

4.3 s/100
mL/sq. in.
Borosilicate Glass, Card Mount
Fast
Grade GF/A is used for high-efficiency general purpose laboratory filtration, including water pollution monitoring of effluents, for filtration of water, algae and bacteria cultures, foodstuff analyses, protein filtration, and radioimmunoassay of weak β emitters. Recommended for gravimetric determination of airborne particulates, stack sampling, and absorption methods of air pollution monitoring.
37 mm
Disc
1.6 μm
4.3 s/100
mL/sq. in.
GF/A (This data shown may be based on historic Whatman records. As is normal practice, customers should determine and validate for themselves (as necessary) that the products are suitable for their specific application.)
37 mm
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit