Learn More
DWK Life Sciences Wheaton™ Boston Round Bottle, Safety Coated, 500mL
Wheaton clear safety coated Boston Round bottles are manufactured from clear soda lime glass that conforms to USP Type III requirements.
Manufacturer:  DWK Life Sciences W217851
 View more versions of this product
Catalog No. 02-927-149
  Check Availability
Add to cart
Description
Description
  • Packaged in convenience packs with caps attached to maintain cleanliness
  • Coated glass bottles provide an added measure of safety
  • Case includes hazardous communication label and tamper evident seal if labeling and/or sealing is required
Specifications
Specifications
500mL
PE Cone
Clear
12/Case
Yes
Wide Mouth
Phenolic
Safety Coated
Type III Borosilicate Glass
Boston Round Bottle
Black