Learn More
DWK Life Sciences Wheaton™ Boston Round Bottle, Safety Coated, Amber, 125mL
Wheaton safety coated Boston Round bottles are made from Wheaton 900 amber glass that conforms to USP Type III requirements. Amber glass is ideal for protecting light-sensitive samples during storage.
Manufacturer:  DWK Life Sciences W217950
 View more versions of this product
Catalog No. 03-084-184
  Check Availability
Add to cart
Description
Description
  • Packaged in convenience packs with caps attached to maintain cleanliness
  • Coated glass bottles provide an added measure of safety
  • Case includes hazardous communication label and tamper evident seal if labeling and/or sealing is required
Specifications
Specifications
125mL
PV
Amber
24/Case
Yes
Wide Mouth
PP
Safety Coated
Type III Borosilicate Glass
Boston Round Bottle
White