Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Wheaton Boston Round Bottle, Safety Coated, Amber, 125mL
Wheaton safety coated Boston Round bottles are made from Wheaton 900 amber glass that conforms to USP Type III requirements. Amber glass is ideal for protecting light-sensitive samples during storage.
Manufacturer: DWK Life Sciences W217953
Description
- Packaged in convenience packs with caps attached to maintain cleanliness
- Coated glass bottles provide an added measure of safety
- Case includes hazardous communication label and tamper evident seal if labeling and/or sealing is required
Specifications
125mL | |
PE Cone | |
Amber | |
24/Case | |
Yes | |
Wide Mouth |
Phenolic | |
Safety Coated | |
Type III Borosilicate Glass | |
Boston Round Bottle | |
Black |
Promotions
-
Save Over 90% on Glassware and Plasticware
Take advantage of amazing deals while they last.: 12-31-2019
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title