Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Wheaton™ Boston Round Bottle, Safety Coated, Amber, 500mL
Description
- Packaged in convenience packs with caps attached to maintain cleanliness
- Coated glass bottles provide an added measure of safety
- Case includes hazardous communication label and tamper evident seal if labeling and/or sealing is required
Specifications
Closure Liner | Polyethylene Cone |
Closure Material | Phenolic |
Product Type | Boston Round Bottle |
Capacity (Metric) | 500 mL |
Coating | Safety Coated |
Color | Amber |
Material | Borosilicate Glass (Type 3) |
Quantity | 12/Case |
Closure Color | Black |
Closure Included | Yes |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction