Learn More
Med Vet InternationalSupplier Diversity Partner BOTTLE,DROPPER,.5 OUNCE,48/CS  DFS Item
Manufacturer:  Med Vet InternationalSupplier Diversity Partner DB.5CS
Amber Plastic Dropper Bottle. These convenient multi-purpose plastic amber bottles feature dropper caps that can be removed to administer treatment or to allow refilling. Ideal for storing liquid medications or other products that must be accurately measured and dispensed precisely with the use of included dropper pipette. Shows ounce and mL markings on bottles and mL markings on dropper. Black rubber bulb. Sealing Type: Dropper cap screw closure. 1/2 Ounce, 48/Case
Catalog No. 50-118-0262
Due to product restrictions, please to purchase or view availability for this product.