Learn More
Microsolv Technology CorporationSupplier Diversity Partner BOTTLE MBL PHSE 20L S/C DFS Item
Manufacturer:  Microsolv Technology CorporationSupplier Diversity Partner 5880222
HPLC mobile phase reservoir bottle. Graduated borosilicate glass. 20L capacity. 45mm screw top plastic coated for safety. Cap & pouring ring included. 1 Each.
Catalog No. 50-014-52643