Sartorius™ Bottled Media for Sartorius Membrane Filtration Units Available on GSA/VA Contract for Federal Government customers only.

Lysine agar media for wild yeasts determination

Manufacturer: Sartorius™ 14143A

 View more versions of this product

Catalog No. 14-555-740

  Check Availability
Add to Cart

    Specifications

    Fill Volume 250mL
    For Use With (Application) Membrane filtration method or for direct incubation, for determination of wild Yeasts
    Format Liquid
    Manufacturing Origin Germany
    Media Type Lysine
    Packaging Bottle
    Quantity 250mL

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit