Learn More
PIP™ Bouton™ Bold Professional™ Safety Glasses
Specifications
Certifications/Compliance Meet ANSI Z87.1-1989 safety standards
View More Specs
Products 3
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
19-088-2057
View Documents
PIP™
25062MB000
N/A Black frame, Clear lens This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
19-088-2062
View Documents
PIP™
25062SB000
N/A Blue frame, Clear lens This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
19-088-2060
View Documents
PIP™
25062MB045
N/A Black frame, TrueView lens This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Sleek and dramatic design in lightweight Resilium™ nylon frames promotes compliance in the workplace. This high-tech style features engineered airflow sideshields on a six-base single-curve lens. The bow-flex bayonet-style temples and Soft-Fit™ nose bridge allow for ergonomic, all-day comfort, while the specially designed lens retention channel holds the lens securely for maximum protection. Impact-resistant polycarbonate lens.
Compliance
Meet ANSI Z87.1-1989 standards.
Specifications
Specifications
Meet ANSI Z87.1-1989 safety standards