Learn More
PIP™ Bouton™ Softsides Cover Chemical Splash Goggles
For chemical splash protection
Manufacturer:  PIP™ 2485190400BX
 View more versions of this product
Catalog No. 19-052559
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Clear polycarbonate lens
  • Soft vinyl body fits over most prescription eyewear and molds comfortably to facial contours
  • Added ribs reinforce body for lens retention and superior impact resistance
  • With elastic headband
  • Hooded vents for indirect ventilation
Order Info
Available in standard or Fogless™ versions.
Certifications
CSA standards.
Compliance
Chemical splash goggle meets or exceeds ANSI™ Z87.1-1989
Specifications
Specifications
Cover Goggles; Over-the-glasses
Safety
Chemical splash protection
Polycarbonate
Clear
ANSI Z87.1-1989, CSA standards
Fits over most prescription eyewear
Fogless