Learn More
PIP™ Bouton™ UFO Mercury Safety Glasses
Manufacturer:  PIP™ 25070MB003
 View more versions of this product
Catalog No. 18-999-570C
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Sleek design combines protection and comfort. Spectacles weigh less than 28g, feature three-way Click-to-Fit™ adjustable temples for easy tip adjustment. Ratch-it-Fit feature lets you adjust the angle of the lens. Frameless wraparound polycarbonate lens provides full peripheral vision, impact resistance, and 100% absorption of harmful UV radiation up to 385nm.
Order Info
Frame available in black; blue; and sporty red, white, and blue. Clear, gray, and mirror lenses available; black frame also comes with amber lens.
Compliance
Spectacles meet ANSI Z87.1-1989 standards.
Specifications
Specifications
Amber
<28.35g
Meet ANSI Z87.1-1989 standards and CE.
Black