Learn More
UbpbioSupplier Diversity Partner BOVINE 20S PROTEASOME, 50UG DFS Item
Manufacturer:  UbpbioSupplier Diversity Partner A1401
Proteasomes/Activators/Subunits – Bovine 20S proteasome (UBPBio catalog# A1400, A1401) - Purified from bovine red blood cells. The 20S proteasome has a barrel – shaped structure arranged as four heptomeric rings of αββα. In eukaryotes, each of α and β ring is composed of seven different proteins. The β1, β2 and β5 subunits have ‘caspase-like’, ‘trypsin-like’ and ‘chymotypsin-like’ activities, respectively.
Catalog No. 50-112-6851