Learn More
Bovine Albumin, RIA Grade, Thermo Scientific™
Used in radioimmuno assay.
Manufacturer: LAR NJ Thermo Scientific J1086818
Description
Suitable for use in radioimmuno assay
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Purity grade: RIA grade
- Assay (electrophoresis): ≥98.0%
- Protein: ≥96.0%
- MDLNumber: MFCD00130384
- CAS: 9048-46-8
Specifications
Lyophilized powder | |
6.8 to 7.2 | |
Bovine blood | |
Plastic bottle |
Albumin | |
50 g | |
4°C | |
Lyophilized powder |
Safety and Handling
EINECSNumber : 232-936-2
Recommended Storage : Keep cold
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only