Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bovine Albumin, RIA Grade, Thermo Scientific™

Used in radioimmuno assay.

$105.20 - $624.00

Specifications

Name Albumin
Source Bovine blood
Format Lyophilized powder
Packaging Plastic bottle
pH Range 6.8 to 7.2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ1086809
View Documents
LAR NJ Thermo Scientific
J1086809
10 g
Each for $105.20
Add to cart
 
AAJ1086822
View Documents
LAR NJ Thermo Scientific
J1086822
100 g
Each for $624.00
Add to cart
 
AAJ1086818
View Documents Promotion Details
LAR NJ Thermo Scientific
J1086818
50 g
Each for $247.00
Add to cart
 
Description

Description

  • Purity grade: RIA grade
  • Assay (electrophoresis): ≥98.0%
  • Protein: ≥96.0%
  • MDLNumber: MFCD00130384
  • CAS: 9048-46-8
Specifications

Specifications

Albumin
Lyophilized powder
6.8 to 7.2
Bovine blood
Plastic bottle
4°C
Safety and Handling

Safety and Handling

EINECSNumber : 232-936-2

Recommended Storage : Keep cold

SDS
Product Certifications

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit