Learn More
BioworldSupplier Diversity Partner BOVINE APROTININ DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220600131
Recombinant Bovine Aprotinin (AP) Source: Maize (Corn) Background: Aprotinin inhibits the activity of several proteolytic enzymes such as chymotrypsin, kallikrein, plasmin and trypsin. It is present in blood and in most tissues, wi...
Catalog No. 50-253-106