Learn More
Raybiotech IncSupplier Diversity Partner Bovine bFGF ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELBBFGF1
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 4.1 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Bovine bFGF.
Catalog No. 50-194-8856