Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Bovine Free Tri-Iodothyronine ELISA Kit (Colorimetric)

High sensitivity and specificity for detection of bovine free Tri-Iodothyronine.

$501.50

Specifications

Product Type ELISA Kit (Colorimetric)
Assay Range 2 to 32pmol/L
Species Bovine
Sensitivity 0.38pmol/L
Sample Type Serum, plasma
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB600141KT View Documents Novus Biologicals™
NBP2600141KIT
1 Kit Each for $501.50
Add to cart
 
Description

Description

The Bovine Free Tri-Iodothyronine ELISA Kit (Colorimetric) measures bovine Free Tri-Iodothyronine in serum, plasma.
Specifications

Specifications

ELISA Kit (Colorimetric)
Bovine
Serum, plasma
For detection of bovine FT3
Free Tri-Iodothyronine
2 to 32pmol/L
0.38pmol/L
RUO
Store the unopened product at 2 to 8°C Protect from light. Do not use past expiration date.
ELISA

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit