Learn More
Bovine IgG, F(ab)2, HRP, Isotype control, Novus Biologicals™
Bovine Isotype Control
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP197224
Catalog No. NBP197224
Add to cart
Description
Description
IgG F(ab)2 antibody specifically detects IgG F(ab)2 in Bovine samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
IgG F(ab)2
HRP
Lyophilized from 0.01 M Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with No Preservative
Bovine
Ion exchange chromatography
RUO
Primary
Reconstitute with 1.0 ml deionized water (or equivalent) to desired concentration.
LYOPH
Restore with 1.0mL of deionized water (or equivalent).
Conjugated
IgG2
1 mg
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase, anti- Bovine IgG, anti-Bovine IgG F(ab)2 and anti-Bovine Serum. No reaction was observed against anti-Bovine IgG F(c) or anti-Pepsin.
Bovine