Learn More
Biomatik Corporation Bovine Interferon Gamma Induced Protein 10kDa (IP10) ELISA Kit
Supplier:  Biomatik Corporation EKU05107
Encompass_Preferred
Bovine Interferon Gamma Induced Protein 10kDa (IP10) ELISA Kit for detecting IP10 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q2KIQ8. Gene ID: 615107. Target Synonyms: CXCL10; C7; IFI10; INP10; SCYB10; Crg2; GIP10; Mob1; Chemokine C-X-C Motif Ligand 10; Small Inducible Cytokine Subfamily B (Cys-X-Cys) Member 10. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8918
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.