Learn More
Raybiotech IncSupplier Diversity Partner Bovine IP-10/CXCL10 ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELBIP101
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.12 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Bovine IP-10.
Catalog No. 50-194-8871