Learn More
Abnova™ Bovine KRT8 native Protein from bovine liver
Used for AP, EIA, SDS-PAGE
Manufacturer:  Abnova™ P5347
Catalog No. 89-950-254
Add to cart
Description
Description
Specifications
Specifications
Native protein purified from bovine liver
Lyophilized
281269
KRT8 (Bovine) Native Protein
≥95% by SDS-PAGE
Store at 4°C. This product is stable for at least 3 months. After reconstitution with 70 ul ddH2O, store at -20°C. Aliquot to avoid repeated freezing and thawing. Reconstitute: Reconstitute to filaments by mixing equimolar amounts of cytokeratins type 1 and type 2 at concentrations of approximately 0.5 mg/mL, in 9.5 M urea buffer. Protofilaments and filament complexes are obtained by dialyzing the resulting polypeptide solution stepwise to a concentration of 4 M urea and
KRT2-8
KRT8
None
Antibody Production, Enzyme Immunoassay, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized from 30mM Tris/HCl, pH 8.0 (9 M urea, 2mM DTT, 2mM EDTA and 10mM CH6ClN)
52.5kDa
Native protein purified from bovine liver
100 ug
RUO
KRT8
Bovine