Learn More
BioworldSupplier Diversity Partner Bovine Serum Albumin (BSA), Fraction V - Cohn, 1 kg DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220700183
Gemini’s Cohn fraction BSA is made by a modification of the original cold-ethanol plasma fractionation process developed in 1939 by Professor Edwin Cohn. Its characteristics include a minimum 96% albumin, with a pH of 7.0 - 7.4. This product is suitable as a growth nutrient in tissue culture media, for increasing the viscosity and protein levels in certain antibiotic assay media, to bind inhibitors in growth media, and as a molecular-weight standard.SpecificationsShips at ambient.
Catalog No. 50-253-921