Learn More
BioworldSupplier Diversity Partner Bovine Serum Albumin (BSA), Fraction V - Low IgG, 1 kg DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220700213
Low-IgG BSA is a highly purified, lyophilized powder. This product is processed to provide a minimum of 98% albumin and to be essentially free of immunoglobulin. The pH is adjusted to 7.0. It is used as a blocking reagent in EIA, RIA and ELISA systems, where IgG may interfere with the application.SpecificationsShips at ambient.
Catalog No. 50-253-930