Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Novus Biologicals™ Bovine Thyroxine ELISA Kit (Colorimetric)

High sensitivity and specificity for detection of bovine T4.

$602.00

Specifications

Product Type ELISA Kit (Colorimetric)
Assay Range 20 to 320ng/mL
Species Bovine
Sensitivity 20ng/mL
Sample Type serum, plasma, tissue homogenates
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB601591KT View Documents Novus Biologicals™
NBP2601591KIT
1 Kit Each for $602.00
N/A
Description

Description

The Bovine Thyroxine ELISA Kit (Colorimetric) measures bovine Thyroxine in serum, plasma, tissue homogenates.
Specifications

Specifications

ELISA Kit (Colorimetric)
Bovine
serum, plasma, tissue homogenates
RUO
For detection of bovine T4
Store the unopened product at 2 to 8°C Protect from light. Do not use past expiration date.
Adhesive Strip (For 96 wells), Assay plate, Conjugate, HRP-avidin, Substrate B, Substrate A, Wash Buffer (20 x concentrate), Stop Solution, Standard, Instruction manual
20 to 320ng/mL
20ng/mL
Signal Transduction
50 to 100μL
T4
Thyroxine
ELISA

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit