Learn More
BioworldSupplier Diversity Partner BOVINE TRYPSIN (RECOMBINANT) DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220600181
Recombinant Bovine Trypsin Source: Corn Background: Trypsin is a serine protease that hydrolyses proteins, it is found in the digestive system of numerous vertebrates. Trypsin is produced as the inactive proenzyme trpsinogen in the pan...
Catalog No. 50-253-112