Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Novus Biologicals™ Bovine Vitamin A ELISA Kit (Colorimetric)

High sensitivity and specificity for detection of bovine Vitamin A.

$732.50

Specifications

Product Type ELISA Kit (Colorimetric)
Assay Range 6.25 to 100ng/mL
Species Bovine
Sensitivity 4.672ng/mL
Sample Type serum, plasma, tissue homogenates
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB601881KT View Documents Novus Biologicals™
NBP2601881KIT
1 Kit Each for $732.50
N/A
Description

Description

The Bovine Vitamin A ELISA Kit (Colorimetric) measures bovine Vitamin A in serum, plasma, tissue homogenates.
Specifications

Specifications

ELISA Kit (Colorimetric)
Bovine
serum, plasma, tissue homogenates
RUO
For detection of bovine VA
Vitamin A
ELISA
6.25 to 100ng/mL
4.672ng/mL
Lipid and Metabolism
50 to 100μL
Store the unopened product at 2 to 8°C Protect from light. Do not use past expiration date.
Adhesive Strip (For 96 wells), Assay plate, HRP-conjugate (100 x concentrate), HRP-conjugate Diluent, Wash Buffer (25 x concentrate), TMB Substrate, Stop Solution, Sample Diluent, Standard (10 x concentrate), Instruction manual

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit