Learn More
TWD Scientific LLC BioX 81-Well Polycarbonate Freezer Box, 3.75 inch, Assorted Colors. Case of 20
Supplier: TWD Scientific LLC BX909381
BioX 3.75 inch 81 well polycarbonate freezer box (dimensions: 132x132x95mm) with removable lid. Boxes/lids have numeric grid for identification purposes. Boxes come in assorted colors (red, yellow, green, blue and white). 20/case, price/case
This item is not returnable.
View return policy