Learn More
GSC Go Science Crazy BOYLE'S Law Deluxe Apparatus Products for Science Education
Used to study the basics of Boyle's Law, as well as estimating the atmospheric pressure. It provides a simple way to approach the experimental side of physics and to introduce at sight the concepts of a vacuum, pressure, and density.
Manufacturer:  GSC Go Science Crazy BOYLAWDLX
Catalog No. S24657
Add to cart
Description
Description
Tube is filled with heavy oil and connected to a closed reservoir, and allows one to study the atmospheric substance in the expansion container. By raising or lowering the other tube end, compression or distillation of the gas is obtained.
Specifications
Specifications
Study the concepts of a vacuum, pressure, and density.
BOYLE'S Law Deluxe Apparatus
Education