Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BPGM Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 171731AP

Catalog No. 50-173-1419


Add to cart

Description

Description

BPGM belongs to the phosphoglycerate mutase family, BPG-dependent PGAM subfamily. It plays a major role in regulating hemoglobin oxygen affinity as a consequence of controlling 2, 3-BPG concentration. It can also catalyze the reaction of EC 5.4.2.1 (mutase) and EC 3.1.3.13 (phosphatase), but with a reduced activity.
Specifications

Specifications

BPGM
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P07738, P15327
2,3 bisphosphoglycerate mutase, Bisphosphoglycerate mutase, BPG dependent PGAM, BPGM
Rabbit
IgG
150 μL
-20°C
Primary
12183, 296973, 669
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
BPGM
Liquid
BPGM
BPGM Fusion Protein Ag10731
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit