Learn More
bPiDDB, Tocris
Orthosteric nAChR antagonist
Manufacturer:  R&D Systems 3221/50
Quantity 50mg
 View more versions of this product
Catalog No. 32-215-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>98%
514.38
bPiDDB
C24H38Br2N2
1, 1'-(1, 12-Dodecanediyl)bis[3-methylpyridinium] dibromide
Specifications
50mg
>98%
514.38
bPiDDB
525596-66-1
C24H38Br2N2
1, 1'-(1, 12-Dodecanediyl)bis[3-methylpyridinium] dibromide
Safety and Handling
Recommended Storage : Desiccate at Room Temperature