Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BPIFA3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549470

Catalog No. PA549470

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

BPIFA3
Polyclonal
PBS with 0.09% sodium azide
Q9BQP9, Q9D9J8
BPI fold-containing family A member 3, Short palate, lung and nasal epithelium carcinoma-associated protein 3, BPIFA3, C20orf71, SPLUNC3
Rabbit
IgG
80 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
128861, 73388
Western Blot
Unconjugated
BPIFA3
Liquid
BPIFA3
KLH conjugated synthetic peptide between 150-185 amino acids from the Central region of human BPIFA3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit