Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BPIFB1 Mouse anti-Mouse, Biotin, Clone: 1F11E3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237435B

Catalog No. NBP237435B

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

BPIFB1
1F11E3
Western Blot, ELISA
Purified
BPIFB1
Purified recombinant fragment of mouse Lplunc1 (AA: 248-475) expressed in E. Coli.
Protein A or G purified
Store at 4C in the dark.
Monoclonal
Mouse
Western Blot, ELISA
Biotin
PBS with 0.05% Sodium Azide
bA49G10.6, chromosome 20 open reading frame 114, dJ1187J4.1, long palate, lung and nasal epithelium carcinoma associated 1, long palate, lung and nasal epithelium carcinoma-associated protein 1, LPLUNC1, MGC14597, VEMSGP, Von Ebner minor salivary gland protein
Mouse
IgG1
0.1 ml
Primary
92747
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit