Learn More
BPIFB1 Mouse anti-Mouse, DyLight 650, Clone: 1F11E3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237435C
Catalog No. NBP237435C
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
BPIFB1
1F11E3
Western Blot, ELISA
Purified
BPIFB1
Purified recombinant fragment of mouse Lplunc1 (AA: 248-475) expressed in E. Coli.
Protein A or G purified
Store at 4C in the dark.
Monoclonal
Mouse
ELISA, Western Blot
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
bA49G10.6, chromosome 20 open reading frame 114, dJ1187J4.1, long palate, lung and nasal epithelium carcinoma associated 1, long palate, lung and nasal epithelium carcinoma-associated protein 1, LPLUNC1, MGC14597, VEMSGP, Von Ebner minor salivary gland protein
Mouse
IgG1
0.1 ml
Primary
92747