Tocris™ BPIPP

Non-competitive GC inhibitor. Also AC inhibitor

Manufacturer: Tocris™ 3635/50

 View more versions of this product

Catalog No. 36-355-0R

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors BPIPP
    Quantity 50mg
    Purity >99
    Molecular Weight 450.28
    Product Type BPIPP

    Non-competitive guanylyl cyclase (GC) and adenylyl cyclase (AC) inhibitor. Downregulates cAMP and cGMP synthesis, and suppresses cGMP accumulation in a variety of cell lines (IC50 = 3.4 - 11.2µM). Inhibits GC-stimulated Cl- transport in vitro and suppresses toxin-induced intestinal fluid accumulation in vivo.

    • Chemical Name: 5-(3-Bromophenyl)-5,11-dihydro-1,3-dimethyl-1H-indeno[2',1':5,6]pyrido[2,3-d]pyrimidine-2,4,6(3H)-trione
    • Formula: C22H16BrN3O3
    • CAS Number: 325746-94-9
    • Solubility: Soluble to 2mM in DMSO
    • Purity: >99

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit