Learn More
Tocris Bioscience™ BPTES Available on GSA/VA Contract for Federal Government customers only.
Glutaminase Inhibitors
$156.40 - $658.50
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
530110 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5301/10
10mg Each for $156.40
Add to cart
530150 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5301/50
Each for $658.50
Add to cart
Description
Description
Allosteric glutaminase (GLS1) inhibitor (IC50 = 3.3 μM). Selective for GLS1 over GLS2 and γ-glutamyl transpeptidase. Induces cell death of P493 human lymphoma B cells in vitro and reduces tumor volume of P493 cell xenografts in mice.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Safety and Handling
Safety and Handling
Recommended Storage : Store at −20°C