Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brachyury Mouse anti-Human, PE, Clone: 1H9A2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237434PE

Catalog No. NBP237434PE

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Brachyury
1H9A2
Flow Cytometry
Purified
T
Purified recombinant fragment of human T (AA: 257-309 ) expressed in E. Coli.
Protein A or G purified
Hematopoietic Stem Cell Markers, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary
6862
Flow Cytometry
PE
PBS with 0.05% Sodium Azide
brachyury protein, MGC104817, Protein T, T brachyury (mouse) homolog, T brachyury homolog, T, brachyury homolog (mouse), TF, TFT
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit