Learn More
Braco Manufacturing Premoistened PPE Cleaning Towelettes
Sanitize personal protection equipment to help prevent cross-contamination
Manufacturer:  Braco Manufacturing ST100DN
Catalog No. 19-161-915
Add to cart
Description
Description
  • For cleaning rubber or latex facepieces, mouthpieces, respirators and more
  • Premoistened with special formula that effectively kills bacteria and removes perspiration and body oils
  • Alcohol free—eliminates worries about “drying out,” typical of alcohol-based cleaners
Order Info
Individually foil wrapped. Packed in ready-to-use dispenser pack with built-in eye hook for easy wall hanging.
Specifications
Specifications
Premoistened Ppe Cleaning Towelettes
Cleaning silicone and plastic facepieces, mouth pieces, respirator, hardhat liners