Learn More
Bradley™ Frost Proof Wall-Mounted Eyewash Unit
Hand operated by a large, highly visible stainless-steel push handle
Manufacturer:  Bradley™ S19220HFP18
Catalog No. 19-807-542
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: Universal identification sign and inspection tag
Description
Description
  • Stainless-steel eyewash receptor
  • Operates quickly by a stainless-steel push flag handle
  • Integral safe, steady water flow in the sprayhead assembly
  • Exceeds minimum water flow of 3 gpm at 30psi
  • BradTech™ corrosion-resistant yellow coating galvanized stee
Compliance
UL classified to meet ANSI Z358.1
Warranty and Services
One year
Specifications
Specifications
Wall mount eyewash fixture
ANSI Z358.1-2009
3 gpm at 30psi
Wall
Safety
Yellow
Stainless steel eyewash bowl, chrome plated brass sprayhead with twin soft flow eyewash head and protective sprayhead cover, NPT stay open chrome plated brass ball valve, galvanized steel, yellow coating pipe and fitting, heavy gauge sand cast Aluminum protected wall bracket, NPT water supply, universal identification sign, inspection tag
30 to 90psi
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
This plumbing fixture is not intended to dispense water for human consumption through drinking or for preparation of food or beverages.