Bradley™ Halo™ Eyewash with SpinTec™ Drench Shower

The Bradley Halo™ Eyewash with SpinTec™ Drench Shower is a space saving fixture equipped with shower providing superior washdown and even spray pattern.

Manufacturer: Bradley™ S19314EW

 View more versions of this product

Catalog No. 19-120-580

  Check Availability
Add to Cart

Disclaimers:

Not intended to dispense water for human consumption through drinking or for preparation of food or beverages.


    Specifications

    Product Type Halo Eyewash
    Flow Rate 0.4 - 20 gpm at 30psi
    For Use With (Application) Full body relief from contaminants
    Includes Plastic SpinTec™ shower head, Plastic bowl, Galvanized Steel, yellow coating pipe and fitting, shower/eye/facewash stay open ball valve, universal identification sign, inspection tag
    Mounting Free standing
    Pressure Range 30 to 90psi
    Activation Stainless Steel push handle
    Certifications/Compliance ANSI Z358.1-2009, EN 15154-1, EN 15154-2, UL
    Color Yellow

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit