Learn More
Bradley™ Halo™ Wall-Mount Eyewash
Save space and and delivers effective eye washdown coverage and spray pattern with the Bradley Halo™ Wall-Mount Eyewash.
Manufacturer:  Bradley™ S19224PDC
 View more versions of this product
Catalog No. 19-120-573
  Check Availability
Add to cart
Disclaimers:
Not intended to dispense water for human consumption through drinking or for preparation of food or beverages.
Specifications Description & Specifications
Specifications
Halo Eyewash
ANSI Z358.1-2009, ADA, EN 15154-1, EN 15154-2, UL
13.25 in.
0.4 gpm at 30psi
14.62 in.
Plastic bowl, Galvanized Steel, yellow coating pipe and fitting, eyewash stay open Chrome Plated Brass ball valve, universal identification sign, inspection tag, Plastic dust cover
30 to 90psi
39.37cm
Stainless Steel push handle
Yellow
33.65cm
Instant relief to eyes from harmful particulates or chemicals
37.13cm
Wall
15.5 in.