Learn More
Bradley™ OneStep™ Personal Eyewash Station
Sleek profile allows for versatile placement in labs where space is at a premium
Alert:
User should proceed to an eyewash station capable of a full 15-minute flush after use of eyewash bottle.
Products 2
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
19-130-1982
Accessories View Documents
Bradley™
S19926
N/A Eyewash Station, One-step, 16 oz. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
19-130-1986
View Documents
Bradley™
S19924
N/A Replacement, One-step, 16 oz. This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Provides immediate flushing of the eyes when a plumbed or portable eyewash is not readily available.
  • Unique, patent-pending design provides one-step activation for bottled eyewash
  • One-hand release with easy-to-open bottle—no twisting or turning required
  • Securely mounts to wall with two screws for quick access
  • Bright yellow UV-protected polyethylene dust cover keeps bottle clean
  • Bottle includes trilingual written and visual instructions for use
Order Info
Bottles printed with expiration date. Shelf life of three years.
Compliance
Complies with ANSI™ Z358.1-2004 as a personal wash unit only. (Does not meet the criteria of plumbed or self-contained eyewash equipment.)