Learn More
Bradley™ OneStep™ Personal Eyewash Station
Sleek profile allows for versatile placement in labs where space is at a premium
Manufacturer:  Bradley Corporation S19924
 View more versions of this product
Catalog No. 19-130-1986
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
User should proceed to an eyewash station capable of a full 15-minute flush after use of eyewash bottle.